RIE156330636 Rieke Metals, Inc.

Ethyl 5-(2,3,4-trimethoxyphenyl)-5-oxovalerate

  • MDL number MFCD09801965
  • Cas Number : 854859-30-6
  • Molecular Weight : 310.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7713C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7713C
Availability details for
Quantity
  • Availability
  • Add to Cart