RIE156330637 Rieke Metals, Inc.

Ethyl 6-(2,3,4-trimethoxyphenyl)-6-oxohexanoate

  • MDL number MFCD09801966
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7713D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7713D
Availability details for
Quantity
  • Availability
  • Add to Cart