RIE156330640 Rieke Metals, Inc.

Ethyl 4-(4-n-Butylphenyl)-4-oxobutanoate

  • MDL number MFCD02261389
  • Cas Number : 115199-55-8
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7714B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7714B
Availability details for
Quantity
  • Availability
  • Add to Cart