RIE156330645 Rieke Metals, Inc.

Ethyl 4-(4-n-Propylphenyl)-4-oxobutanoate

  • MDL number MFCD02261379
  • Cas Number : 57821-80-4
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7715B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7715B
Availability details for
Quantity
  • Availability
  • Add to Cart