RIE156330650 Rieke Metals, Inc.

Ethyl 4-[2-(Methylthio)phenyl]-4-oxobutanoate

  • MDL number MFCD09801969
  • Molecular Weight : 252.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7716B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7716B
Availability details for
Quantity
  • Availability
  • Add to Cart