RIE156330652 Rieke Metals, Inc.

Ethyl 4-[4-(Ethylthio)phenyl]-4-oxobutanoate

  • MDL number MFCD07782909
  • Cas Number : 7028-71-9
  • Molecular Weight : 266.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7717B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7717B
Availability details for
Quantity
  • Availability
  • Add to Cart