RIE156330653 Rieke Metals, Inc.

Ethyl 5-[4-(Ethylthio)phenyl]-5-oxovalerate

  • MDL number MFCD07782910
  • Molecular Weight : 280.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7717C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7717C
Availability details for
Quantity
  • Availability
  • Add to Cart