RIE156330654 Rieke Metals, Inc.

Ethyl 4-(3-Ethoxyphenyl)-4-oxobutanoate

  • MDL number MFCD09801971
  • Cas Number : 905592-32-7
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7718B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7718B
Availability details for
Quantity
  • Availability
  • Add to Cart