RIE156330655 Rieke Metals, Inc.

Ethyl 5-(3-Ethoxyphenyl)-5-oxovalerate

  • MDL number MFCD09801972
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7718C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7718C
Availability details for
Quantity
  • Availability
  • Add to Cart