RIE156330657 Rieke Metals, Inc.

Ethyl 5-(3-Methyl-2-thienyl)-5-oxovalerate

  • MDL number MFCD09801974
  • Molecular Weight : 240.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7719C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7719C
Availability details for
Quantity
  • Availability
  • Add to Cart