RIE156330658 Rieke Metals, Inc.

Ethyl 4-(5-Chloro-2-thienyl)-4-oxobutanoate

  • MDL number MFCD07782905
  • Cas Number : 473694-31-4
  • Molecular Weight : 246.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7720B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7720B
Availability details for
Quantity
  • Availability
  • Add to Cart