RIE156330661 Rieke Metals, Inc.

Ethyl 5-(3-Bromo-2-thienyl)-5-oxovalerate

  • MDL number MFCD07782896
  • Molecular Weight : 305.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7721C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7721C
Availability details for
Quantity
  • Availability
  • Add to Cart