RIE156330663 Rieke Metals, Inc.

Ethyl 5-[3,4-(methylenedioxy)phenyl]-5-oxovalerate

  • MDL number MFCD07782914
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7722C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7722C
Availability details for
Quantity
  • Availability
  • Add to Cart