RIE156330666 Rieke Metals, Inc.

Ethyl 4-(4-tert-Butyl-2,6-dimethylphenyl)-4-oxobutanoate

  • MDL number MFCD09801975
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7724B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7724B
Availability details for
Quantity
  • Availability
  • Add to Cart