RIE156330671 Rieke Metals, Inc.

ethyl 5-oxo-5-phenylvalerate

  • MDL number MFCD00032371
  • Cas Number : 73172-56-2
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7509C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7509C
Availability details for
Quantity
  • Availability
  • Add to Cart