RIE156330673 Rieke Metals, Inc.

Ethyl 6-oxo-6-phenylhexanoate

  • MDL number MFCD00032372
  • Cas Number : 4248-25-3
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7509D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7509D
Availability details for
Quantity
  • Availability
  • Add to Cart