RIE156330674 Rieke Metals, Inc.

Ethyl 7-oxo-7-phenylheptanoate

  • MDL number MFCD00672059
  • Cas Number : 112665-41-5
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7509E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7509E
Availability details for
Quantity
  • Availability
  • Add to Cart