RIE156330675 Rieke Metals, Inc.

ethyl 8-oxo-8-phenyloctanoate

  • MDL number MFCD01320194
  • Cas Number : 103187-95-7
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7509F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7509F
Availability details for
Quantity
  • Availability
  • Add to Cart