RIE156330678 Rieke Metals, Inc.

5-Oxo-heptanoic acid

  • MDL number MFCD01320159
  • Cas Number : 3637-13-6
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7087C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7087C
Availability details for
Quantity
  • Availability
  • Add to Cart