RIE156330679 Rieke Metals, Inc.

Ethyl 4-(4-ethoxyphenyl)-4-oxobutyrate

  • MDL number MFCD00568430
  • Cas Number : 339289-35-9
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7655B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7655B
Availability details for
Quantity
  • Availability
  • Add to Cart