RIE156330680 Rieke Metals, Inc.

Ethyl 7-(4-methoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD01311624
  • Cas Number : 122115-54-2
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7511E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7511E
Availability details for
Quantity
  • Availability
  • Add to Cart