RIE156330681 Rieke Metals, Inc.

ethyl 8-(4-methoxyphenyl)-8-oxooctanoate

  • MDL number MFCD01320195
  • Cas Number : 362669-41-8
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7511F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7511F
Availability details for
Quantity
  • Availability
  • Add to Cart