RIE156330688 Rieke Metals, Inc.

4-(4-hexyloxyphenyl)-4-oxobutyric acid

  • MDL number MFCD00100660
  • Cas Number : 64779-14-2
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7160B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7160B
Availability details for
Quantity
  • Availability
  • Add to Cart