RIE156330690 Rieke Metals, Inc.

6-Oxo-octanoic acid

  • MDL number MFCD01320160
  • Cas Number : 4233-57-2
  • Molecular Weight : 158.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7087D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7087D
Availability details for
Quantity
  • Availability
  • Add to Cart