RIE156330691 Rieke Metals, Inc.

5-(2,4-Dichlorophenyl)-5-oxovaleric acid

  • MDL number MFCD02260931
  • Cas Number : 172167-99-6
  • Molecular Weight : 261.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7143C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7143C
Availability details for
Quantity
  • Availability
  • Add to Cart