RIE156330695 Rieke Metals, Inc.

4-Oxo-4-(2-pyridyl)butyric acid

  • MDL number MFCD01320175
  • Cas Number : 5768-27-4
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7096B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7096B
Availability details for
Quantity
  • Availability
  • Add to Cart