RIE156330699 Rieke Metals, Inc.

4-Oxo-4-(3-pyridyl)butyric acid

  • MDL number MFCD00870737
  • Cas Number : 4192-31-8
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7097B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7097B
Availability details for
Quantity
  • Availability
  • Add to Cart