RIE156330700 Rieke Metals, Inc.

5-Methyl-4-oxohexanoic acid

  • MDL number MFCD00154360
  • Cas Number : 41654-04-0
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7131B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7131B
Availability details for
Quantity
  • Availability
  • Add to Cart