RIE156330702 Rieke Metals, Inc.

4-(4-Ethylphenyl)-4-oxobutyric acid

  • MDL number MFCD00173766
  • Cas Number : 49594-75-4
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7146B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7146B
Availability details for
Quantity
  • Availability
  • Add to Cart