RIE156330705 Rieke Metals, Inc.

4-(2,4-Dimethoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD01098010
  • Cas Number : 14617-06-2
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7166B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7166B
Availability details for
Quantity
  • Availability
  • Add to Cart