RIE156330707 Rieke Metals, Inc.

Ethyl 4-oxohexanoate

  • MDL number MFCD00068986
  • Cas Number : 3249-33-0
  • Molecular Weight : 158.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7587B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7587B
Availability details for
Quantity
  • Availability
  • Add to Cart