RIE156330708 Rieke Metals, Inc.

7-Methyl-6-oxooctanoic acid

  • MDL number MFCD02260881
  • Cas Number : 59210-01-4
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7131D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7131D
Availability details for
Quantity
  • Availability
  • Add to Cart