RIE156330710 Rieke Metals, Inc.

5-(4-n-Butylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD06201673
  • Cas Number : 845781-46-6
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7270

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7270
Availability details for
Quantity
  • Availability
  • Add to Cart