RIE156330711 Rieke Metals, Inc.

4-Oxooctanoic acid

  • MDL number MFCD00520681
  • Cas Number : 4316-44-3
  • Molecular Weight : 158.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7088B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7088B
Availability details for
Quantity
  • Availability
  • Add to Cart