RIE156330712 Rieke Metals, Inc.

Ethyl 4-oxooctanoate

  • MDL number MFCD00520679
  • Cas Number : 37174-96-2
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7588B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7588B
Availability details for
Quantity
  • Availability
  • Add to Cart