RIE156330713 Rieke Metals, Inc.

Ethyl 5-oxononanoate

  • MDL number MFCD01320341
  • Cas Number : 24071-99-6
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7588C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7588C
Availability details for
Quantity
  • Availability
  • Add to Cart