RIE156330714 Rieke Metals, Inc.

Ethyl 6-oxodecanoate

  • MDL number MFCD01320342
  • Cas Number : 4144-61-0
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7588D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7588D
Availability details for
Quantity
  • Availability
  • Add to Cart