RIE156330715 Rieke Metals, Inc.

Ethyl 7-oxoundecanoate

  • MDL number MFCD01311673
  • Cas Number : 227953-80-2
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7588E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7588E
Availability details for
Quantity
  • Availability
  • Add to Cart