RIE156330719 Rieke Metals, Inc.

4-Oxo-4-(4-pyridyl)butyric acid

  • MDL number MFCD01320178
  • Cas Number : 5693-75-4
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7098B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7098B
Availability details for
Quantity
  • Availability
  • Add to Cart