RIE156330721 Rieke Metals, Inc.

Ethyl 4-(4-methylphenyl)-4-oxobutyrate

  • MDL number MFCD00266564
  • Cas Number : 6942-61-6
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7531B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7531B
Availability details for
Quantity
  • Availability
  • Add to Cart