RIE156330722 Rieke Metals, Inc.

5-(2,5-Dimethoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD00667603
  • Cas Number : 63467-20-9
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7167C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7167C
Availability details for
Quantity
  • Availability
  • Add to Cart