RIE156330723 Rieke Metals, Inc.

6-(2,5-Dimethoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD02261034
  • Cas Number : 79381-16-1
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7167D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7167D
Availability details for
Quantity
  • Availability
  • Add to Cart