RIE156330726 Rieke Metals, Inc.

5-(4-isopropylphenyl)-5-oxovaleric acid

  • MDL number MFCD02260953
  • Cas Number : 18847-18-2
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7148C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7148C
Availability details for
Quantity
  • Availability
  • Add to Cart