RIE156330732 Rieke Metals, Inc.

5-Oxononanoic acid

  • MDL number MFCD01320162
  • Cas Number : 3637-15-8
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7088C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7088C
Availability details for
Quantity
  • Availability
  • Add to Cart