RIE156330738 Rieke Metals, Inc.

3-(1-Oxo-2-phenylethyl)benzoic acid

  • MDL number MFCD01003946
  • Cas Number : 423151-69-3
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7100

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7100
Availability details for
Quantity
  • Availability
  • Add to Cart