RIE156330740 Rieke Metals, Inc.

Ethyl 4-oxo-4-(4-pyridyl)butyrate

  • MDL number MFCD01320358
  • Cas Number : 25370-46-1
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7598B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7598B
Availability details for
Quantity
  • Availability
  • Add to Cart