RIE156330743 Rieke Metals, Inc.

4-(4-t-butylphenyl)-4-oxobutyric acid

  • MDL number MFCD02260961
  • Cas Number : 35288-08-5
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7150B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7150B
Availability details for
Quantity
  • Availability
  • Add to Cart