RIE156330747 Rieke Metals, Inc.

5-(3,4-Dimethoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD00094346
  • Cas Number : 4378-55-6
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7169C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7169C
Availability details for
Quantity
  • Availability
  • Add to Cart