RIE156330750 Rieke Metals, Inc.

Ethyl 3-(2-oxopropyl)benzoate

  • MDL number MFCD02261281
  • Cas Number : 73013-49-7
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7609

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7609
Availability details for
Quantity
  • Availability
  • Add to Cart