RIE156330753 Rieke Metals, Inc.

Ethyl 3-acetylbenzoate

  • MDL number MFCD02261283
  • Cas Number : 37847-24-8
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7612

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7612
Availability details for
Quantity
  • Availability
  • Add to Cart